Anti-TNFSF13B Rabbit Polyclonal Antibody

Supplier: Proteintech
11862-1-AP-150UL
10096-140EA 402 USD
10096-140
Anti-TNFSF13B Rabbit Polyclonal Antibody
Antibodies
TNFSF13B, also named as BAFF, BLYS, TALL1, TNFSF20, ZTNF4 and CD257, is a cytokine that binds to TNFRSF13B/TACI and TNFRSF17/BCMA. This cytokine is expressed in B cell lineage cells, and acts as a potent B cell activator. It has been also shown to play an important role in the proliferation and differentiation of B cells. This antibody recognizes all the forms of TNFSF13B such as soluble form (17 kDa), membrane form (32-35 kDa) and glycosylation form (45-52 kDa).

Western Blot: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: TNFSF13B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR