Anti-RAMP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11861-1-AP-150UL
10093-300EA 402 USD
10093-300
Anti-RAMP3 Rabbit Polyclonal Antibody
Antibodies
The receptor activity-modifying proteins (RAMPs) and the calcitonin receptor-like receptor (CRLR) are both required to generate adrenomedullin (AM) and calcitonin gene-related peptide (CGRP) receptors. During the process, RAMP3 transport the glycosylated 58-kD CGRPR to the cell surface. The formation of CRLR-RAMP heterodimer is essential for the proper cell surface targeting and pharmacological characteristics of both CGRP and AM receptors.

Western Blot: Mouse Heart Tissue, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RAMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR