Anti-SIRPG Rabbit Polyclonal Antibody

11857-1-AP
10094-406EA 356.38 USD
10094-406
Anti-SIRPG Rabbit Polyclonal Antibody
Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: SIRPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR