Anti-CASP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
11856-1-AP-150UL
10084-090EA 402 USD
10084-090
Anti-CASP4 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SGC-7901 Cells, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CASP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR