Anti-Cofilin 2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11848-1-AP-150UL
10084-962EA 459.7 USD
10084-962
Anti-Cofilin 2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: Cofilin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR