Anti-SRPX2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11845-1-AP-150UL
10095-188EA 459.7 USD
10095-188
Anti-SRPX2 Rabbit Polyclonal Antibody
Antibodies
SRPX2 (sushi-repeat-containing protein, X-linked 2) is a secreted protein expressed in neurons of the human adult brain, including the rolandic area . Firstly described as sushi-repeat protein up-regulated in leukemia (SPRUL) in leukemia cells with dysregulated expression at the transcriptional level, SRPX2 has been recently shown as a multifunctional protein. SRPX2 is a ligand for uPAR, the urokinase-type plasminogen activator (uPA) receptor . It is involved in seizure disorders, angiogenesis and cellular adhesion . The involvement of SRPX2 in disorders of language cortex and cognition suggests it has an important role in the perisylvian region critical for language and cognitive development .

Western Blot: Human Placenta Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: SRPX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR