Anti-Langerin Rabbit Polyclonal Antibody

Supplier: Proteintech
11841-1-AP-150UL
10089-504EA 402 USD
10089-504
Anti-Langerin Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human skin Tissue, 1:200-1:2000

Type: Primary
Antigen: Langerin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR