Anti-Surfactant protein D Rabbit Polyclonal Antibody

Supplier: Proteintech
11839-1-AP-150UL
10094-476EA 402 USD
10094-476
Anti-Surfactant protein D Rabbit Polyclonal Antibody
Antibodies
IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: Surfactant protein D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR