Anti-MLF2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11835-1-AP-150UL
10090-266EA 459.7 USD
10090-266
Anti-MLF2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: MLF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR