Anti-UBE2K Rabbit Polyclonal Antibody

Supplier: Proteintech
11834-3-AP-150UL
10096-644EA 459.7 USD
10096-644
Anti-UBE2K Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: UBE2K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR