CIB1(Calcium and integrin-binding protein 1) is also named as CIB, KIP, PRKDCIP. It is an EF-hand-containing protein that binds multiple effector proteins, including the platelet αIIbβ3 integrin and several serine/threonine kinases and potentially modulates their function. It regulates platelet aggregation in hemostasis through a specific interaction with the alphaIIb cytoplasmic domain of platelet integrin alphaIIbbeta3.
Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200
Type: Primary
Antigen: CIB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: