Anti-PSG1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11809-1-AP-150UL
10092-878EA 402 USD
10092-878
Anti-PSG1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: PSG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR