Anti-S100 P Rabbit Polyclonal Antibody

Supplier: Proteintech
11803-1-AP-150UL
10094-100EA 402 USD
10094-100
Anti-S100 P Rabbit Polyclonal Antibody
Antibodies
S100P protein is a relatively small (95 amino acid) isoform of the S100 protein family that was first isolated from human placenta. Overexpression of S100P has been detected in several cancers such as breast, colon, prostate, pancreatic and lung carcinomas, and the protein has been functionally implicated in carcinogenic processes. S100P protein is widely expressed in both normal and neoplastic tissues. It clearly shows ectopic expression in some cancers. Based on the high expression in certain tumors, S100P could represent a potential target for novel diagnostic and therapeutic applications.

IHC: Human lung cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP:N/A, 1:500-1:5000; Western Blot: HeLa Cells, 1:500-1:5000; FC: HepG2 Cells, N/A

Type: Primary
Antigen: S100 P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR