Anti-TOMM20 Rabbit Polyclonal Antibody

Supplier: Proteintech
11802-1-AP-150UL
10096-180EA 537.7 USD
10096-180
Anti-TOMM20 Rabbit Polyclonal Antibody
Antibodies
TOMM20, also named as KIAA0016, belongs to the Tom20 family. It is a central component of the receptor complex responsible for the recognition and translocation of cytosolically synthesized mitochondrial preproteins. Together with TOM22, TOMM20 functions as the transit peptide receptor at the surface of the mitochondrion outer membrane and facilitates the movement of preproteins into the TOM40 translocation pore. TOMM20 is characterized as major docking receptors to mediate the recognition by different mechanisms.

Western Blot: Mouse Brain Tissue, 1:1000-1:10000; IHC: Human gliomas Tissue, 1:20-1:200; IP: HEK-293 Cells, 1:1000-1:10000; FC: HeLa Cells, N/A

Type: Primary
Antigen: TOMM20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR