Anti-NA14 Rabbit Polyclonal Antibody

Supplier: Proteintech
11797-1-AP-150UL
10090-792EA 402 USD
10090-792
Anti-NA14 Rabbit Polyclonal Antibody
Antibodies
SSNA1, also named as NA14 and DIP13, may be an important adaptor to target spastin activity at the centrosome. SSNA1 is a 14 kDa protetin which locates the basal bodies and flagella of human sperm cells and the centrosome of cells.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Colon cancer, 1:20-1:200

Type: Primary
Antigen: NA14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR