Anti-KYNU Rabbit Polyclonal Antibody

Supplier: Proteintech
11796-1-AP-150UL
10089-406EA 402 USD
10089-406
Anti-KYNU Rabbit Polyclonal Antibody
Antibodies
KYNU(kynureninase) is also named as L-kynurenine hydrolase and belongs to the kynureninase family. It is a pyridoxal-5'-phosphate-(pyridoxal-P)-depe11dent enzyme that catalyses the cleavage of L-kynurenine and L-3-hydroxykynurenine into anthranilic and 3-hydroxyanthranilic acids, respectively. In SDS-PAGE, it can detect a 48 kDa band lower than the preidicted molecular weight of 52 kDa due to limited proteolysis of the enzyme which might occur during handling of the protein. This protein is a 95-kD kD homodimer predominantly located in the cytoplasm. It has 2 isoforms produced by alternative splicing.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: KYNU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR