Anti-TREM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11791-1-AP-150UL
10096-304EA 402 USD
10096-304
Anti-TREM1 Rabbit Polyclonal Antibody
Antibodies
Triggering receptor expressed on myeloid cells 1 (TREM1, also known as CD354) is a member of the immunoglobulin superfamily, which is expressed on neutrophils and monocytes. TREM1 plays a role in the innate immune system. TREM1 amplifies neutrophil and monocyte-mediated inflammatory responses triggered by bacterial and fungal infections by stimulating release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers.

Western Blot:PC-3 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; FC: HeLa Cells, N/A

Type: Primary
Antigen: TREM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR