Anti-CCL11 Rabbit Polyclonal Antibody

Supplier: Proteintech
11786-1-AP-150UL
10086-552EA 402 USD
10086-552
Anti-CCL11 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Heart Tissue, 1:1000-1:10000

Type: Primary
Antigen: CCL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR