Anti-LY96 Rabbit Polyclonal Antibody

Supplier: Proteintech
11784-1-AP-150UL
10089-426EA 402 USD
10089-426
Anti-LY96 Rabbit Polyclonal Antibody
Antibodies
Western Blot:U-937 Cells, 1:200-1:2000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: LY96
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR