Anti-PTPN22 Rabbit Polyclonal Antibody

Supplier: Proteintech
11783-1-AP-150UL
10093-062EA 402 USD
10093-062
Anti-PTPN22 Rabbit Polyclonal Antibody
Antibodies
PTPN22(Tyrosine-protein phosphatase non-receptor type 22) is also named as LyP,PEP,PTPN8.It belongs to the protein-tyrosine phosphatase family and non-receptor class 4 subfamily.It acts as negative regulator of T-cell receptor (TCR) signaling.Defects in PTPN22 are a cause of susceptibility to systemic lupus erythematosus (SLE).It has 5 isoforms produced by alternative splicing.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200

Type: Primary
Antigen: PTPN22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR