Anti-IGKV1-5 Rabbit Polyclonal Antibody

Supplier: Proteintech
11781-1-AP-150UL
10088-674EA 459.7 USD
10088-674
Anti-IGKV1-5 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A549 Cells, 1:500-1:5000

Type: Primary
Antigen: IGKV1-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR