Anti-NAMPT Rabbit Polyclonal Antibody

Supplier: Proteintech
11776-1-AP-150UL
10096-980EA 599.76 USD
10096-980
Anti-NAMPT Rabbit Polyclonal Antibody
Antibodies
Nicotinamide phosphoribosyltransferase(NAMPT) has two usual synonyms termed Visfatin and PBEF. Its primary role is to catalyze the condensation of nicotinamide with 5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, an intermediate in the biosynthesis of NAD, which is the rate limiting component in the mammalian NAD biosynthesis pathway. NAMPT is localized in cytoplasm and expressed in large amounts in bone marrow, liver tissue, and muscle tissues. NAMPT inhibits neutrophil apoptosis in experimental inflammation and clinical sepsis. NAMPT levels are altered in plasma of patients with type 2 diabetes mellitus (T2DM), and it is now evidenced that NAMPT may plays a role in lipid metabolism.

Western Blot: Mouse Heart Tissue, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IP: Mouse Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: NAMPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR