Anti-NFIL3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11773-1-AP-150UL
10091-080EA 402 USD
10091-080
Anti-NFIL3 Rabbit Polyclonal Antibody
Antibodies
NFIL3, also named as E4BP4 and IL3BP1, belongs to the bZIP family and NFIL3 subfamily. It represses transcription from promoters with activating transcription factor (ATF) sites. NFIL3 activates transcription from the interleukin-3 promoter in T-cells. It competes for the same consensus-binding site with PAR DNA-binding factors (DBP, HLF and TEF). It is a component of the circadian clock that acts as a negative regulator for the circadian expression of PER2 oscillation in the cell-autonomous core clock. NFIL3 protects pro-B cells from programmed cell death. It is a key regulator of TH2 responses.

Western Blot: HepG2 Cells, 1:200-1:2000; IP: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: NFIL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR