Anti-SFRS6 Rabbit Polyclonal Antibody

Supplier: Proteintech
11772-1-AP
10094-460EA 440.73 USD
10094-460
Anti-SFRS6 Rabbit Polyclonal Antibody
Antibodies
SR proteins have a characteristic C-terminal Ser/Arg-rich repeat (RS domain) of variable length and constitute a family of highly conserved nuclear phosphoproteins that can function as both essential and alternative pre-mRNA splicing factors . SRFS6 has a pivotal role in the TF exon 5 definition, an event essential for flTF biosynthesis. Besides, up- or down regulation of SRSF6 alters the splicing of several tumor suppressors and oncogenes to generate the oncogenic isoforms and reduce the tumor suppressive isoforms .

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: SFRS6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR