Anti-CYP4B1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11771-1-AP-150UL
10085-540EA 402 USD
10085-540
Anti-CYP4B1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse lung Tissue, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200; IP: Mouse lung Tissue, 1:200-1:2000

Type: Primary
Antigen: CYP4B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR