Anti-SEPT8 Rabbit Polyclonal Antibody

Supplier: Proteintech
11769-1-AP-150UL
10094-326EA 402 USD
10094-326
Anti-SEPT8 Rabbit Polyclonal Antibody
Antibodies
SEPT8, also named as KIAA0202, is a member of the highly conserved septin family. Septins are 40- to 60-kD GTPases that assemble as filamentous scaffolds. They are involved in the organization of submembranous structures, in neuronal polarity, and in vesicle trafficking. It may play a role in platelet secretion. SEPT8 is about 50-58 kDa in WB detection.

Western Blot:K-562 Cells, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200

Type: Primary
Antigen: SEPT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR