Anti-PANK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11768-1-AP-150UL
10091-782EA 402 USD
10091-782
Anti-PANK1 Rabbit Polyclonal Antibody
Antibodies
PANK1(Pantothenate kinase 1) is also named as PANk and belongs to the type II pantothenate kinase family. The PANK1 protein, which catalyzes the rate-limiting step of coenzyme A biosynthesis, is also upregulated by p53. It also catalyzes the cytosolic phosphorylation of pantothenate (vitamin B5) and derivatives. This protein has 4 isoforms produced by alternative splicing with the molecular weight of 64 kDa, 42 kDa, 36kDa and 45 kDa,respectively.

Western Blot: Raji Cells, 1:200-1:1000

Type: Primary
Antigen: PANK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR