Anti-CLPS Rabbit Polyclonal Antibody

11766-1-AP
10084-878EA 349.28 USD
10084-878
Anti-CLPS Rabbit Polyclonal Antibody
Western Blot:BxPC-3 Cells, 1:200-1:2000

Type: Primary
Antigen: CLPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR