Anti-HNRNPC Rabbit Polyclonal Antibody

Supplier: Proteintech
11760-1-AP-150UL
10088-376EA 402 USD
10088-376
Anti-HNRNPC Rabbit Polyclonal Antibody
Antibodies
HNRNPC, also named as Heterogeneous nuclear ribonucleoproteins C1/C2, is a 306 amino acid protein, which contains 1 RRM (RNA recognition motif) domain and belongs to the RRM HNRPC family. HNRNPC localizes in the nucleus. It binds pre-mRNA and nucleates the assembly of 40S hnRNP particles. HNRNPC may play a role in the early steps of spliceosome assembly and pre-mRNA splicing. HNRNPC can act as a tetramer and is involved in the assembly of 40S hnRNP particles and plays a role with CSDE1 in internal initiation of translation during mitosis.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human Lymphoma Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; FC: HeLa Cells, N/A

Type: Primary
Antigen: HNRNPC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR