Anti-ZNF395 Rabbit Polyclonal Antibody

Supplier: Proteintech
11759-1-AP-150UL
10097-348EA 459.7 USD
10097-348
Anti-ZNF395 Rabbit Polyclonal Antibody
Antibodies
ZNF395 is a transcription factors binding to a 7bp GC rich sequence which resides in triplicate at intervals of 13bp within and proximal to the -20bp direct repeat sequences of the Htt promoter. It can bind to regulatory regions in papillomaviruses (PV) and subsequently called the protein papillomavirus binding factor (PBF) . It contains conserved regions CR1, CR2 and CR3, a domain rich in serines and prolines and have the potential to form a zinc-finger structure, and the C-terminal CR3 is responsible for DNA-binding .

Western Blot: Human Brain Tissue, 1:200-1:1000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: ZNF395
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR