Anti-NFIA Rabbit Polyclonal Antibody

Supplier: Proteintech
11750-1-AP-150UL
10091-076EA 402 USD
10091-076
Anti-NFIA Rabbit Polyclonal Antibody
Antibodies
The NFI (nuclear factor I) family consists of four members in vertebrates (NFI-A, NFI-B, NFI-C and NFI-X), and the four NFI genes are expressed in unique patterns during mouse embryogenesis and in the adult. Four isoforms of NFIA were found in human and they play various roles in DNA replication, DNA-dependent transcritpion via their DNA binding property. Multiple residues of NFIA can be phosphorylated resulting in mild shifts of its practical molecular weight. Recent finding also revealed its neuroprotective function in NMDA-induced neuronal damage. Catalog# 11750-1-AP is a rabbit polyclonal antibody raised against N-terminal of human original NFIA.

Western Blot:A431 Cells, 1:200-1:2000; IHC: Human prostate cancer, 1:10-1:100

Type: Primary
Antigen: NFIA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR