CACYBP(Calcyclin-binding protein) is also named as S100A6BP, SIP. It is a new target 30 kDa protein that can interact with S100 proteins, including S100A6, S100A1, S100B, and S100P, but not to S100A4, calbindin D, parvalbumin or calmodulin. It may be involved in calcium-dependent ubiquitination and subsequent proteasomal degradation of target proteins and probably serves as a molecular bridge in ubiquitin E3 complexe. It has 2 isoforms produced by alternative splicing and can exsit as a dimer.
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; FC: HeLa Cells, N/A
Type: Primary
Antigen: CACYBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat