Anti-SPRR3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11742-1-AP-150UL
10095-124EA 402 USD
10095-124
Anti-SPRR3 Rabbit Polyclonal Antibody
Antibodies
SPRR3, also named as Esophagin, Cornifin beta and SPRC, belongs to the cornifin (SPRR) family. It is cross-linked envelope protein of keratinocytes. Increased expression of SPRR3 appears to be involved in colorectal tumorigenesis. SPRR3 is a marker for terminal squamous cell differentiation.a potential novel therapeutic target for less advanced stages of Breast Cancer.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200; IP:COLO 320 Cells, 1:200-1:2000

Type: Primary
Antigen: SPRR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR