Anti-ASB8 Rabbit Polyclonal Antibody

Supplier: Proteintech
11735-1-AP-150UL
10083-180EA 402 USD
10083-180
Anti-ASB8 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Heart Tissue, 1:200-1:1000; IHC: Human Colon cancer, 1:20-1:200

Type: Primary
Antigen: ASB8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR