Anti-YPEL5 Rabbit Polyclonal Antibody

Supplier: Proteintech
11730-1-AP-150UL
10097-218EA 459.7 USD
10097-218
Anti-YPEL5 Rabbit Polyclonal Antibody
Antibodies
YPEL5(Protein yippee-like 5) belongs to the yippee family and is involved in a certain cell division-related function.During cell cycle, YPEL5 protein is detected at different subcellular localizations; at interphase, it is located in the nucleus and centrosome, then it changes location sequentially to spindle poles, mitotic spindle, and spindle midzone during mitosis, and finally transferrs to midbody at cytokinesis.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Prostate Cancer Tissue, 1:10-1:100

Type: Primary
Antigen: YPEL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR