Anti-MASPIN Rabbit Polyclonal Antibody

Supplier: Proteintech
11722-1-AP-150UL
10089-646EA 402 USD
10089-646
Anti-MASPIN Rabbit Polyclonal Antibody
Antibodies
SERPINB5, also named as PI-5, Maspin and Serpin B5, belongs to the serpin family and Ov-serpin subfamily. It is a tumor suppressor. It blocks the growth, invasion, and metastatic properties of mammary tumors. As it does not undergo the S (stressed) to R (relaxed) conformational transition characteristic of active serpins, SERPINB5 exhibits no serine protease inhibitory activity.

Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: MASPIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR