Anti-MNAT1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11719-1-AP-150UL
10090-322EA 459.7 USD
10090-322
Anti-MNAT1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:2000; IF:A375 Cells, 1:10-1:100

Type: Primary
Antigen: MNAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR