Anti-GAR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11711-1-AP-150UL
10087-390EA 402 USD
10087-390
Anti-GAR1 Rabbit Polyclonal Antibody
Antibodies
GAR1, other named NOLA1, is a subunit of H/ACA and telomerase. Thought is not required for H/ACA protein assembly, GAR1 is necessary for ribosome biogenesis and telomere. H/ACA involves in class specify the sites of uridine-to-pseudouridine. It contains a core domain that flanked by glycine- and arginine-rich(GAR) domains. GAR1 also required for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transciptase(TERT) holoenzyme.

Western Blot:A375 Cells, 1:200-1:2000; IHC: Human skin cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP:A375 Cells, 1:200-1:1000

Type: Primary
Antigen: GAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR