Anti-MRPL23 Rabbit Polyclonal Antibody

Supplier: Proteintech
11706-1-AP-150UL
10090-434EA 402 USD
10090-434
Anti-MRPL23 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A2780 Cells, 1:200-1:2000; IF:PC-3 Cells, 1:10-1:100

Type: Primary
Antigen: MRPL23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR