Anti-MCM5 Rabbit Polyclonal Antibody

Supplier: Proteintech
11703-1-AP-150UL
10090-048EA 402 USD
10090-048
Anti-MCM5 Rabbit Polyclonal Antibody
Antibodies
Minichromosome maintenance, S. Cerevisiae, homolog of 5(MCM5), one component of the complex of MCM2-7, which is the putative replicative essential for once per cell cycle DNA replication initiation and elongation in eukaryotic cells. The MCM complex form a hexameric ring through the interaction surfaces of two neighboring subunits.

Western Blot:Jurkat Cells, 1:1000-1:10000; IHC: Human Lymphoma, 1:20-1:200

Type: Primary
Antigen: MCM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR