Anti-TPST2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11699-1-AP
10095-904EA 370.14 USD
10095-904
Anti-TPST2 Rabbit Polyclonal Antibody
Western Blot: Human Liver Tissue, 1:200-1:2000; IHC: Human pancreas cancer, 1:20-1:200

Type: Primary
Antigen: TPST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR