Anti-Importin 13 Rabbit Polyclonal Antibody

Supplier: Proteintech
11696-2-AP-150UL
10088-952EA 459.7 USD
10088-952
Anti-Importin 13 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human gliomas Tissue, 1:20-1:200; IF: HeLa Cells, 1:20-1:200

Type: Primary
Antigen: Importin 13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR