Anti-ARL5B Rabbit Polyclonal Antibody

Supplier: Proteintech
11694-1-AP-150UL
10082-668EA 402 USD
10082-668
Anti-ARL5B Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: ARL5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR