Anti-GNG2 Rabbit Polyclonal Antibody

Supplier: Proteintech
11693-1-AP-150UL
10087-702EA 459.7 USD
10087-702
Anti-GNG2 Rabbit Polyclonal Antibody
Antibodies
Gγ2 subunit (Gng2/GNG2) is one of subunits of the Gβγ-dimer composing heterotrimeric G protein with a Gα-subunit. Heterotrimeric G protein has been reported to be involved in various biological activities including cell proliferation, differentiation, invasion and angiogenesis. It is expressed in a range of foetal tissues as well as adult testis, adrenal gland, brain, white blood cells and lung.

Western Blot: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: GNG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR