Anti-TAX1BP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11692-1-AP-150UL
10095-618EA 459.7 USD
10095-618
Anti-TAX1BP3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse ovary Tissue, 1:500-1:5000; IHC: Human small intestine Tissue, 1:20-1:200

Type: Primary
Antigen: TAX1BP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR