Anti-DYNLT3 Rabbit Polyclonal Antibody

Supplier: Proteintech
11687-1-AP-150UL
10085-594EA 459.7 USD
10085-594
Anti-DYNLT3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:1000-1:10000; IHC: Human ovary tumor, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: DYNLT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR