Anti-ARPP19 Rabbit Polyclonal Antibody

Supplier: Proteintech
11678-1-AP-150UL
10083-152EA 459.7 USD
10083-152
Anti-ARPP19 Rabbit Polyclonal Antibody
Antibodies
ARPP-19, a 19-kDa cAMP-regulated phosphoprotein, plays a role in regulating mitosis. Initiation and maintenance of mitosis require the activation of protein kinase cyclin B–Cdc2 and the inhibition of protein phosphatase 2A (PP2A). When phosphorylated by protein kinase Greatwall (Gwl), ARPP-19 associate with and inhibit PP2A, thus promoting mitotic entry. ARPP-19 may be an important link between nerve growth factor (NGF) signaling and post-transcriptional control of neuronal gene expression such as GAP-43.

Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:200-1:2000; IF:A375 Cells, 1:10-1:100

Type: Primary
Antigen: ARPP19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR