Anti-PDLIM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11674-1-AP-150UL
10091-840EA 459.7 USD
10091-840
Anti-PDLIM1 Rabbit Polyclonal Antibody
Antibodies
PDLIM1, also named as CLIM1, CLP-36, CLP36, hCLIM1 and Elfin, is a member of the actinin-associated LIM protein (ALP)/enigma protein family, plays a role in neurite outgrowth in the peripheral nervous system. It is a cytoskeletal protein that may act as an adapter that brings other proteins (like kinases) to the cytoskeleton. PDLIM1 and PALLD formed a complex in the dorsal root ganglion (DRG) and PDLIM1 and PALLD were colocalized in the neurites and cell bodies of primary DRG neurons. PDLIM1 may interact with PALLD to influence neurite outgrowth during sciatic nerve regeneration.

Western Blot: Mouse lung Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: PDLIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR