Anti-MCHR1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11656-2-AP-150UL
10090-032EA 402 USD
10090-032
Anti-MCHR1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SH-SY5Y Cells, 1:500-1:5000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: MCHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR