Anti-SLC22A18 Rabbit Polyclonal Antibody

Supplier: Proteintech
11646-2-AP-150UL
10094-724EA 402 USD
10094-724
Anti-SLC22A18 Rabbit Polyclonal Antibody
Antibodies
SLC22A18 is a member of the family of polyspecific organic-cation transporters that localize at the apical membrane surface of renal proximal tubules. SLC22A18 is expressed at high levels in the kidney, liver, colon and in fetal renal proximal tubules and present at lower levels in heart, brain and lung. Defects in SLC22A18 are associated with breast and lung cancer and are the cause of rhabdomyosarcoma type 1, a malignant tumor derived from striated muscle. More recently, SLC22A18 has been shown to functions as a tumor suppressor in glioma and represents a candidate biomarker for long-term survival in this disease.

Western Blot: Mouse Brain Tissue, 1:1000-1:10000

Type: Primary
Antigen: SLC22A18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR